Z285323
Glas-Col® Series TM reaction flask mantle
for flask size, 2000 mL, for ACE flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
AC/DC input
115 V
feature
CSA approved
for flask size 2000 mL
manufacturer/tradename
Glas-Col 100B-TM566
availability
not available in EU
output
450 watts
Looking for similar products? Visit Product Comparison Guide
General description
Provides even heat for reaction kettles – ideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size. Reaction flask not included.
- Not CE compliant
Other Notes
Must be used with power controller.
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service