Z275727
Nalgene® stirring rods
polypropylene
Synonym(s):
plastic stirring rod, stirring paddle
All Photos(1)
About This Item
UNSPSC Code:
41103800
NACRES:
NB.35
Recommended Products
material
polypropylene
packaging
pkg of 12 ea
manufacturer/tradename
Nalgene 6169-0010
L
9 3/4 in.
General description
Combines the functions of a stirring rod and a rubber policeman. Paddles at each end, 5/8 in. W.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries