Skip to Content
Merck
All Photos(1)

Key Documents

WHA10535097

Whatman® qualitative filter paper, Grade CF 12

sheets, W × L 210 mm × 297 mm, pack of 100

Synonym(s):

filter paper, whatman filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104929
NACRES:
NB.24

material

sheets

feature

Grade CF 12

packaging

pack of 100

manufacturer/tradename

Whatman 10535097
Whatman Article No. 28416911 (US reference)

W × L

210 mm × 297 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This high-quality filter paper is ideal for applications requiring efficient filtration and separation of fine particles, ensuring reliable results in laboratory settings.

Features and Benefits

  • Grade CF 12
  • 210 mm × 297 mm

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service