TEM-CF300CUUL
Ultrathin Carbon Film Supported Copper Square Mesh
size 300 mesh, box of 50
Synonym(s):
CARBON SQUARE MESH - CU 300 MESH UL 50/BX
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
packaging
box of 50
size
300 mesh
Related Categories
General description
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Copper grid, 300 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)