Skip to Content
Merck
All Photos(1)

Key Documents

T70602

Sigma-Aldrich

3,4,5-Trimethoxyphenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH2CO2H
CAS Number:
Molecular Weight:
226.23
Beilstein:
2697844
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

117-120 °C (lit.)

SMILES string

COc1cc(CC(O)=O)cc(OC)c1OC

InChI

1S/C11H14O5/c1-14-8-4-7(6-10(12)13)5-9(15-2)11(8)16-3/h4-5H,6H2,1-3H3,(H,12,13)

InChI key

DDSJXCGGOXKGSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmet Şenocak et al.
Dalton transactions (Cambridge, England : 2003), 47(29), 9617-9626 (2018-07-04)
The synthesis and characterization of new hybrid materials based on reduced graphene oxide (rGO) or single walled carbon nanotubes (SWCNTs) covalently functionalized by 4,4'-difluoro-8-(4-propynyloxy)-phenyl-1,3,5,7-tetramethyl-4-bora-3a,4a-diaza-s-indacene (BODIPY) (2) or 7-(prop-2-yn-1-yloxy)-3-(3',4',5'-trimethoxyphenyl)-coumarin (4) as light harvesting groups have been described. The organic solar cell
M I Donnelly et al.
Journal of bacteriology, 147(2), 477-481 (1981-08-01)
When grown at the expense of 3,4,5-trimethoxyphenylacetic acid, a species of Arthrobacter readily oxidized 3,4-dihydroxy-5-methoxyphenylacetic acid, but other structurally related aromatic acids were oxidized only slowly. Cell extracts contained a dioxygenase for 3,4-dihydroxy-5-methoxyphenylacetate, and the corresponding trihydroxy acid, which was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service