SYNF284060X
Synthware™ fritted gas dispersion tube with 90 degree angle
disc diam. 40 mm, porosity: extra coarse, stem O.D. × L 8 mm × 150 mm
Synonym(s):
Synthware F284060X, glass gas dispersion tube
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
colorless tube
glass tube
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F284060X
disc diam.
40 mm
stem O.D. × L
8 mm × 150 mm
Related Categories
General description
The 90° angle permits bubbles to be spread over a large area. Stem is 8 mm O.D. × 150 Length; XC stands for extra coarse porosity
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)