NORS405001
Bruker Match™ tubes
volume 310 μL
Synonym(s):
precision machined NMR tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
Recommended Products
material
colorless tube
feature
neck thread push fit
packaging
pack of 5 ea
manufacturer/tradename
Norell S-4.0-500-1
volume
310 μL
wall thickness
0.79 mm
color
blue cap
Looking for similar products? Visit Product Comparison Guide
General description
Ultra-precision machined NMR tubes for the Bruker MATCH system. Caps are color-coded by size, order separately.
Recommended sample volumes: Use 3.0 mm to 5 mm O.D. tubes for 5 mm RT probe, and 1.0 mm to 2.5 mm O.D. tubes for 3 mm Cryo-Probe. 1.0 mm size not recommended for dedicated 1 mm Bruker probe.
Recommended sample volumes: Use 3.0 mm to 5 mm O.D. tubes for 5 mm RT probe, and 1.0 mm to 2.5 mm O.D. tubes for 3 mm Cryo-Probe. 1.0 mm size not recommended for dedicated 1 mm Bruker probe.
Legal Information
Match is a trademark of Bruker Biospin Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service