Skip to Content
Merck
All Photos(1)

Key Documents

M1125

Sigma-Aldrich

L-(−)-Malic acid sodium salt

95-100% (enzymatic)

Synonym(s):

L-Hydroxybutanedioic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H5O5Na
CAS Number:
Molecular Weight:
156.07
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Assay

95-100% (enzymatic)

SMILES string

[Na].O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5.Na/c5-2(4(8)9)1-3(6)7;/h2,5H,1H2,(H,6,7)(H,8,9);/q;+1/p-1/t2-;/m0./s1

InChI key

DOJOZCIMYABYPO-DKWTVANSSA-M

General description

L-Malic acid is the naturally occurring isomer of malic acid, found mainly in sour and unripe fruits.

Application

L-(-)-Malic acid sodium salt may be used to prepare standard solution during the analysis of wine samples using peroxidase based biosensors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxidase based biosensors for the selective determination of D, L-lactic acid and L-malic acid in wines.
Mazzei F, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 87(1), 81-86 (2007)
A Bolekova et al.
European journal of histochemistry : EJH, 56(4), e50-e50 (2013-01-31)
The purpose of this study was to investigate the effect of prenatal administration of retinoic acid (RA) on the development of nicotinamide adenine dinucleotide diaphorase (NADPH-d) positive structures in the rat Meibomian glands. One mg/kg of RA was applied to
Mc Murry JE
Organic Chemistry , Biological Approach (2016)
A Bolekova et al.
European journal of histochemistry : EJH, 55(3), e23-e23 (2011-11-11)
The purpose of this study was to analyze the nicotinamide adenine dinucleotide phosphate - diaphorase (NADPH-d) activity in the rat jejunum after a mesenteric ischemia/reperfusion injury. Nitric oxide, synthetised from L-arginine by the enzyme nitric oxide synthase, is a nonadrenergic
Alexandria M H Lesicko et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(43), 11037-11050 (2016-11-01)
The lateral cortex of the inferior colliculus receives information from both auditory and somatosensory structures and is thought to play a role in multisensory integration. Previous studies in the rat have shown that this nucleus contains a series of distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service