GF62639163
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5
foil, 150 x 150mm, 0.030mm thickness, annealed, not light tested (nlt)
Synonym(s):
TI040300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
not light tested (nlt)
form
annealed (
quarter hard)
foil (
quarter hard)
manufacturer/tradename
Goodfellow 626-391-63
size × thickness
150 mm × 150 mm × 0.030 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service