Skip to Content
Merck
All Photos(1)

Key Documents

GF43061352

Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3

foil, 50 x 50mm, 0.9mm thickness, annealed, mirror polished on both sides

Synonym(s):

FF210376

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

annealed (mirror polished on both sides)
foil

manufacturer/tradename

Goodfellow 430-613-52

size × thickness

50 mm × 50 mm × 0.9 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service