GF29017380
Alumina/Silica/Boria
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 100 m, filament diameter 0.011mm
Synonym(s):
AJ625710
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
fibers
manufacturer/tradename
Goodfellow 290-173-80
L
100 m
diam.
0.011 mm , filament diameter
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service