CLS710310
Pyrex® volumetric pipette, to contain, to deliver, Class A
red, volume 10 mL, accuracy: 0.02 mL
Synonym(s):
Corning® 7103
About This Item
Recommended Products
material
glass
feature
class A
graduation
packaging
case of 6 ea
pack of 6 ea
manufacturer/tradename
Corning 7103-10
volume measuring range
10 mL, accuracy: 0.02 mL
L
429 mm
capacity
10 mL
color
red
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® volumetric pipettes offer high precision measurement of liquids.
- Calibrated "To Contain" (bottom line) and "To Deliver" (top line) to specifications for Class A.
- Color-coded with colored graduations for easy identification.
- Calibration lines enameled onto the glass to ensure volume accuracy.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service