Key Documents
CLS44602L
Pyrex® baffled trypsinizing flask, with beaded neck
capacity 1,500 mL
Synonym(s):
Corning® 4460
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
material
borosilicate glass
narrow-mouth
sterility
non-sterile
feature
closure type Cotton plugs
packaging
case of 2
manufacturer/tradename
Corning 4460-2L
capacity
1,500 mL
General description
Flasks are used for converting homogenous tissue samples into cell suspensions by digestion of connective proteins. Heavy beaded neck accepts cotton plugs. Four extra-deep baffles enhance vigorous agitation.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)