Skip to Content
Merck
All Photos(1)

Key Documents

BR781912

BRAND® microplate BRANDplates®, inertGrade

size 96 wells, size 330 μL, flat bottom, white, with transparent bottom

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, inertGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

flat bottom
polystyrene
white wells

sterility

sterile

feature

lid
skirt (F-bottom, transparent bottom)

packaging

pack of 5 ea

manufacturer/tradename

BRAND 781912

size

330 μL
96 wells

working volume

330 μL

color

white, with transparent bottom

Looking for similar products? Visit Product Comparison Guide

General description

For cultivation of suspension cell lines.Especially suited for cell cultures, when adhesion is not desired.Optimized surface characteristics reduce cell adhesion and protein adsorption, minimizing enzyme and cellular activation.Inhibits early differentiation of stem cells.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service