BR781315
BRAND® PCR tubes, strips of 8
Synonym(s):
BIO-CERT, PCR, PCR Tube Strips, PCR Tubes, Tube Strips, Tubes
About This Item
Recommended Products
manufacturer/tradename
BRAND
technique(s)
PCR: suitable
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | BR781317 | BR781333 | BR781318 |
---|---|---|---|
technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable |
General description
Application
Features and Benefits
- High flexibility with a wide range of cap types and film strips
- Reduced evaporation losses thanks to tightly closing cap strips
- Rapid heat transfer through thin vessel walls
- Numbered wells for clear identification
- Free of: DNA, RNA, DNase, RNase, pyrogens, PCR inhibitors
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service