BR38206
BRAND® volumetric flasks with 3 marks
capacity 25 mL, joint: 10/19, polypropylene stopper
Synonym(s):
BLAUBRAND®, DAkkS, Volumetric flasks
About This Item
Recommended Products
material
borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS neck10/19
joint: 10/19
feature
class A DE-M marking, DIN EN ISO 1042. Calibrated to contain (TC, In). Incl. DAkkS-certificate.
packaging
pack of 1 ea
manufacturer/tradename
BRAND 38206
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- BRAND® volumetric flasks with 3 marks are used for the accurate measurement of instruments for precision analyses.
- Calibrated to contain (TC, In).
- The volumetric flask is used to check the functioning of a dispenser. The mark in the middle shows the nominal volume, the upper and lower marks show the error limits. If the error limit is exceeded even with repeated measurements, the dispenser is out of specification.
- The DAkkS-calibrated volumetric flask does not replace the gravimetric test of the monitoring of measuring instruments according to ISO 8655.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service