BAH131820815
Clavies® autoclave bags
clear, pack of 100 ea
Synonym(s):
sterilzation bag, transparent autoclave bags
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
clear
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art H13182-0815
width
200 mm
Related Categories
General description
These transparent autoclave bags are made of 0.05mm (2 mil) thick polypropylene for discarding used Petri dishes, membrane filters, multi-well cell culture plates, cell culture flasks, culture plates, pipettes, and more.
Features and Benefits
- Three largest sizes hold 65, 200 and 240 standard Petri dishes respectively
- Steam autoclavable at 135°C (275°F)
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)