BAF247390001
SP Bel-Art® Poxygrid Glove Dispenser
epoxy-coated (wire), to hold, Single Box Holder- holds 1x glove box
Synonym(s):
Glove box holder, glove dispenser
All Photos(2)
About This Item
material
epoxy-coated (wire)
feature
to hold Single Box Holder- holds 1x glove box
to hold Single Box Holder
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24739-0001
H × L × width
8.25 in. (210 mm) × 5.5 in. (140 mm) × 4.25 in. (108 mm)
Related Categories
General description
Keep disposable gloves handy with these welded steel wire racks. Select rack to hold one, two or three boxes of disposable gloves, wipes or tissues.
Features and Benefits
- Glove box drops into an individual retaining compartment and lifts out easily for replacement
- Retaining compartment internal dimensions are 12.7cmW x 9.5cmD (5 x 3¾")
- Designed for mounting to a vertical surface
- Durable, blue epoxy coated racks are steam autoclavable at 121°C (250°F)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP