Skip to Content
Merck
All Photos(1)

Key Documents

BAF131653748

Super strength biohazard disposal bags

width × H × thickness 37 in. (940 mm) × 48 in. (1220 mm) × 0.05 mm (2 mil), pack of 100 ea, red

Synonym(s):

biohaz waste bag, biohazard disposal bags, waste bag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.

material

red

packaging

pack of 100 ea

manufacturer/tradename

Bel-Art F13165-3748

capacity

151.4-189.3 L (40-50 gal)

width × H × thickness

37 in. (940 mm) × 48 in. (1220 mm) × 0.05 mm (2 mil)

Looking for similar products? Visit Product Comparison Guide

General description

Protect against dangerous tears and leaks with extra strong, high impact polypropylene biohazard disposal bags.

Features and Benefits


Red bags printed with standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • Extra heavy-duty; 0.05mm (2 mil) thick blended polymer
  • Autoclavable to 135°C (275°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • Extra large sizes available, call for quote


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service