Merck
All Photos(7)
Key Documents
BAB609105000
SP Bel-Art® Durac® Bluetooth Thermometer
temp. range at -50-300 °C, stainless steel probe
Synonym(s):
Stainless Steel Probe Bluetooth Thermometer
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
material
stainless steel probe
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60910-5000
range of application measuring range
-50-300 °C, resolution: 1 °F/C (±1°C from -20/130°C; ±2°C from -50/-21°C and 131/200°C; otherwise ±4°C)
parameter
temp. range at -50-300 °C
cable L
1 m , 3.3"
probe L
150 mm , 6"
battery power
2 AAA
General description
Remotely monitor temperature by broadcasting readings to any Apple or Android smartphone or device with Bluetooth 4.0 or better utilizing the free THERMSmart app.
Features and Benefits
  • Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
  • Alarm sounds to alert user if temperature falls outside of the set parameters Includes countdown timer function
  • Alarm and timer settings savable for reuse
  • LCD screen for on-unit monitoring
  • App can trigger indicator light for unit identification -Indicators include active alarm state and low battery
  • Verification model provides accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
  • Includes Certificate of Compliance indicating accuracy for 4 temperature points and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions
  • Individually serialized, CE marked and RoHS compliant -Batteries included
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP