Skip to Content
Merck
All Photos(1)

Documents

A-LMPAKMM

Sigma-Aldrich

Activation Kit For Multiplex Microspheres

Synonym(s):

AnteoTech Microsphere Activation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115865
NACRES:
NA.22

usage

10 reactions

availability

not available in Japan

storage temp.

2-8°C

General description

AnteoTech identified the following problems that researchers are having with their multiplex assays:
  • Binding difficult antibodies and antigens to microspheres
  • Complex protocols for inexperienced users
  • Lengthy microsphere and protein preparation times prior to protein coupling
  • Reproducibility between batches of protein coupled microspheres
  • Limited storage of ready-to-couple microspheres

From these findings, they developed the AnteoBind Activation Kit For Multiplex Microspheres that is for use with the Luminex xMAP® system and contains the patented AnteoBind Activation Reagent.

With this Activation Kit, you can couple multiple antibodies without extra optimization for each, saving assay development time and reducing the barriers of assay transfer between diverse platforms.

AnteoBind is water based and replaces the dry chemicals you would use with the EDC method. Using AnteoBind instead of NHS/EDC chemistry can save up to 3-4 hours of lab time. Experiments have shown increased stability and functionality for antibodies and a decreased background. AnteoBind improves reproducibility between experiments by providing uniformity between surfaces. You can store your AnteoBind activated microspheres for up to 1 year.

POSTER: A Novel User Friendly Chemistry to Couple Antibodies and Challenging Proteins to Luminex MagPlex Microspheres

Application

Activation and coupling of Luminex Microspheres for Multiplex Immunoassays

Features and Benefits

  • Water based AnteoBind activation and coupling reagent for microspheres
  • Fast assay development time making it easy to move your ELISA assay to the Luminex xMAP® system
  • Compatible with both Luminex MagPlex® and MicroPlex® microspheres
  • Test problematic proteins on xMAP, including those cited as not suitable for the system
  • An alternative to traditional chemistries such as NHS/EDC methods with an easier to use AnteoBind protocol
  • Ease of use, simply pipette AnteoBind Reagent to your chosen microspheres to activate
  • Reduce reagent preparation time by 3-4 hours
  • Increase antibody functionality
  • Low background noise
  • High reproducibility between experiments

Legal Information

Anteobind is a trademark of AnteoTech Ltd
MagPlex is a registered trademark of Luminex Corp
MicroPlex is a registered trademark of Luminex Corp
xMAP is a registered trademark of Luminex Corp

Kit Components Only

Product No.
Description

  • Mix&Go Activation Reagent

  • Coupling Buffer

  • Storage Buffer

  • Low Binding Microcentrifuge Tubes

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Bohm et al.
Vaccine, 35(43), 5883-5889 (2017-09-19)
Worldwide about 1.5 million clinical cases of hepatitis A virus (HAV) infections occur every year and increasingly countries are introducing HAV vaccination into the childhood immunization schedule with a single dose instead of the originally licenced two dose regimen. Diagnosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service