Skip to Content
Merck
All Photos(4)

Key Documents

934704

Sigma-Aldrich

Sodium manganese oxide

greener alternative

Na0.44MnO2, powder, ≥99% trace metals basis

Synonym(s):

Sodium manganite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na0.44MnO2
Molecular Weight:
97.05
UNSPSC Code:
26111700
NACRES:
NA.21

grade

battery grade

Quality Level

description

Capacity: 110 mAh/g
Description/Nominal Voltage: 2.5 V vs Na/Na+

Assay

≥99% trace metals basis

form

powder

composition

Na0.44MnO2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

1-2 μm

density

4.3 g/cm3 (lit.)

application(s)

battery manufacturing

greener alternative category

General description

Sodium manganese oxide is a powdery black substance, which crystallizes in an orthorhombic tunnel-type structure. The structure’s interconnected S-shaped tunnels offer wide diffusion paths for fast and reversible sodium intercalation.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Our high-purity sodium manganese oxide, Na0.44MnO2, powder is suitable and tested for use as a cathode material in Na-ion batteries. Na0.44MnO2 is promising for use in sodium-ion batteries because of its unique tunnel structure, which facilitates excellent cycle and rate performance. For example, our sodium manganese oxide showed a large capacity (experimentally 110 mAh g−1 at 0.1C), even at fast charge rates (80 mAh g-1 at 5C, 72%). In addition, Na0.44MnO2 shows excellent stability with NaClO4 and NaPF6-based organic electrolytes and also aqueous electrolytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent Progress in Electrode Materials for Sodium-Ion Batteries
Kim H, et al.
Advanced Energy Materials, 6, 1600943- 1600943 (2016)
High-Performance Na0.44MnO2 Slabs for Sodium-Ion Batteries Obtained through Urea-Based Solution Combustion Synthesis
Ferrara C, et al.
Lithium-Ion Batteries, 4 (2018)
Recent Advances and Prospects of Cathode Materials for Sodium-Ion Batteries
Xiang X, et al.
Advanced Materials, 27, 5343?5364-5343?5364 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service