Skip to Content
Merck
All Photos(1)

Key Documents

921041

Sigma-Aldrich

Dichlorobis(benzonitrile)platinum(II)

≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl2(C6H5CN)2
CAS Number:
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23

Quality Level

Assay

≥99.9%

SMILES string

[Pt](Cl)(Cl)([NH]#Cc2ccccc2)[NH]#Cc1ccccc1

InChI

1S/2C7H5N.2ClH.Pt/c2*8-6-7-4-2-1-3-5-7;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

WAJRCRIROYMRKA-UHFFFAOYSA-L

Application

Dichlorobis(benzonitrile)platinum(II) is commonly used as a precursor to synthesize organoplatinum (II) complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Isolation, Characterization, and Isomerization of cis- and trans-Bis(benzonitrile)dichloroplatinum(II).
Uchiyama T, et al/
Bulletin of the Chemical Society of Japan, 54(1), 181-185 (1981)
1, 2 polybutadiene complexes with d8 pseudo-square- planar- transition-metal salts based on nickel (II), palladium (II), and platinum (II).
Belfiore L A, et al.
Journal of Polymer Science. Part B, Polymer Physics, 42(12), 2270-2285 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service