Skip to Content
Merck
All Photos(3)

Key Documents

911607

Sigma-Aldrich

[Ir(dFOMeppy)2-(5,5′-dCF3bpy)]PF6

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H22F16IrN4O2P
CAS Number:
Molecular Weight:
1069.75
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

>300 °C

photocatalyst activation

465 nm

SMILES string

[Ir+](c5c(c(cc(c5)F)F)c6ncc(cc6)OC)c3c(c(cc(c3)F)F)c4ncc(cc4)OC.FC(F)(F)c1cnc(cc1)c2ncc(cc2)C(F)(F)F.F[P-](F)(F)(F)(F)F

InChI

1S/C12H6F6N2.2C12H8F2NO.F6P.Ir/c13-11(14,15)7-1-3-9(19-5-7)10-4-2-8(6-20-10)12(16,17)18;2*1-16-9-3-5-12(15-7-9)10-4-2-8(13)6-11(10)14;1-7(2,3,4,5)6;/h1-6H;2*2-3,5-7H,1H3;;/q;;;-1;+1

InChI key

VCHHFLJLUYSBMX-UHFFFAOYSA-N

Application

[Ir(dFOMeppy)2-(5,5′-dCF3bpy)]PF6 is a versatile photocatalyst that can be used for a variety of synthetic transformations including the trifluoromethylation of alkyl bromides.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J P Kornfilt et al.
Journal of the American Chemical Society, 141(17), 6853-6858 (2019-04-16)
Copper oxidative addition into organohalides is a challenging two-electron process. In contrast, formal oxidative addition of copper to Csp2 carbon-bromine bonds can be accomplished by employing latent silyl radicals under photoredox conditions. This novel paradigm for copper oxidative addition has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service