Skip to Content
Merck
All Photos(1)

Key Documents

911402

Sigma-Aldrich

(CyPAd-DalPhos)NiCl(otol)

≥95%

Synonym(s):

8-(2-(Dicyclohexylphosphaneyl)phenyl)-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphaadamantane nickel(II) o-tolyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H49ClNiO3P2
CAS Number:
Molecular Weight:
673.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

193-197 °C

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 Inhalation - Muta. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract,Skin

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-Catalyzed N-Arylation of Cyclopropylamine and Related Ammonium Salts with (Hetero)aryl (Pseudo)halides at Room Temperature
Tassone J P, et al.
ACS Catalysis, 7(9), 6048-6059 (2017)
Preston M MacQueen et al.
Journal of the American Chemical Society, 140(15), 5023-5027 (2018-03-31)
The use of (L)Ni( o-tolyl)Cl precatalysts (L = PAd-DalPhos or CyPAd-DalPhos) enables the C( sp2)-O cross-coupling of primary, secondary, or tertiary aliphatic alcohols with (hetero)aryl electrophiles, including unprecedented examples of such nickel-catalyzed transformations employing (hetero)aryl chlorides, sulfonates, and pivalates. In
Nickel-Catalyzed N-Arylation of Cyclopropylamine and Related Ammonium Salts with (Hetero)aryl (Pseudo)halides at Room Temperature
Tassone J P, et al
ACS Catalysis, 7(9), 6048-6059 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service