Skip to Content
Merck
All Photos(2)

Documents

907014

Sigma-Aldrich

Coolade

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H112N2O31
CAS Number:
Molecular Weight:
1453.61
UNSPSC Code:
51171641
NACRES:
NA.22

form

paste

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

27-30 °C

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry.This product has been enhanced for "Designing Safer Chemicals".
Click here for more information.

Application

Coolade is a low-foaming designer surfactant developed in the Lipshutz Lab tailored to enable organic synthesis in water. When used in aqueous micellar catalytic redutions, Coolade minimizes foaming traditionally resulting from the combination of gas evolution and the use of a surfactant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas R Lee et al.
ChemSusChem (2019-03-20)
Several types of reduction reactions in organic synthesis are performed under aqueous micellar-catalysis conditions (in water at ambient temperature), which produce a significant volume of foam owing to the combination of the surfactant and the presence of gas evolution. The

Articles

Surfactants for efficient reactions in water as a green chemistry alternative.

Surfactants for efficient reactions in water as a green chemistry alternative.

Surfactants for efficient reactions in water as a green chemistry alternative.

Surfactants for efficient reactions in water as a green chemistry alternative.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service