Skip to Content
Merck
All Photos(1)

Key Documents

906549

Sigma-Aldrich

SLAM-VproR-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

SLAM-VproS-13CH3 Kit has13CH3 isotopomer precursor and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For detection of long-range nOes or for the NMR study of large proteins
High incorporation of 13CH3 isotopomer of methyl groups in valine residues without detectable scrambling is achieved using SLAM-VproS-13CH3 kit. This kit has been tested with protein isotopic labeling in E. coli. It enables investigation of supramolecular complex upto 1 MDa in solid-state NMR spectroscopy studies.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific isotopic labeling of methyl groups for NMR spectroscopic studies of high-molecular-weight proteins.
Pierre Gans et al.
Angewandte Chemie (International ed. in English), 49(11), 1958-1962 (2010-02-17)
Rime Kerfah et al.
Current opinion in structural biology, 32, 113-122 (2015-04-17)
Nuclear magnetic resonance (NMR) spectroscopy is a uniquely powerful tool for studying the structure, dynamics and interactions of biomolecules at atomic resolution. In the past 15 years, the development of new isotopic labeling strategies has opened the possibility of exploiting
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service