Skip to Content
Merck
All Photos(2)

Documents

904457

Sigma-Aldrich

Silica

monodisperse, non-porous, 1.0 μm

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41123003
NACRES:
NA.23

form

powder

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging whereas micron sized particles are used in liquid chromatography. We offer highly monodisperse(coefficient of variance <10%) nonporous silica in size varying from 150 nm to 50 μm. The high monodispersity of our particles makes them suitable for aforementioned applications.
Silica monodispersed, non-porous (1 μm) is a micro-sized filler material can be used as a spacer in liquid crystals, high performance liquid chromatography (HPLC), and nanomedicine for biological applications.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-depth characterization of slurry packed capillary columns with 1.0-mm nonporous particles using reversed-phase isocratic ultrahigh-pressure liquid chromatography
Patel KD, et al.
Analytical Chemistry, 76(19), 5777-5786 (2004)
Size control of monodisperse nonporous silica particles by seed particle growth
Zhao B, et al.
Particuology, 9(3), 314-317 (2011)
Practical Examination of a Nonporous Silica Stationary Phase for Reversed-Phase Fast LC Applications
D R Jenke
Journal of Chromatographic Science, 34, 362-362 (1996)
Nonporous silica nanoparticles for nanomedicine application
Tang L and Cheng J
Nano Today, 8(3), 290-312 (2013)
Li Tang et al.
Nano today, 8(3), 290-312 (2013-09-03)
Nanomedicine, the use of nanotechnology for biomedical applications, has potential to change the landscape of the diagnosis and therapy of many diseases. In the past several decades, the advancement in nanotechnology and material science has resulted in a large number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service