Skip to Content
Merck
All Photos(1)

Key Documents

809861

Sigma-Aldrich

Etiocholanolone-2,2,3,4,4-d5 solution

100 μg/mL in methanol, ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D5H25O2
Molecular Weight:
295.47
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

liquid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C([2H])([2H])[C@]([2H])(O)C([2H])([2H])C[C@]4(C)[C@@]3([H])CC[C@@]21C

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13-,14+,15+,16+,18+,19+/m1/s1/i7D2,11D2,13D

InChI key

QGXBDMJGAMFCBF-JHABVYBQSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service