Skip to Content
Merck
All Photos(1)

Key Documents

776157

Sigma-Aldrich

2-(Trifluoromethoxy)ethanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5F3O2
CAS Number:
Molecular Weight:
130.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.323

density

1.334 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCOC(F)(F)F

InChI

1S/C3H5F3O2/c4-3(5,6)8-2-1-7/h7H,1-2H2

InChI key

HZAARXYRNUBDNS-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.9 °F

Flash Point(C)

44.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service