775614
1-Bromo-3-nitrobenzene
97%
Synonym(s):
3-Bromo-1-nitrobenzene, 3-Nitro-1-bromobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01
Beilstein:
2043212
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
crystals
mp
52-55 °C
functional group
bromo
nitro
SMILES string
[O-][N+](=O)c1cccc(Br)c1
InChI
1S/C6H4BrNO2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H
InChI key
FWIROFMBWVMWLB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wenqing Xu et al.
Environmental science & technology, 49(6), 3419-3426 (2015-02-12)
Sorption to black carbons is an important sink for organic contaminants in sediments. Previous research has suggested that black carbons (graphite, activated carbon, and biochar) mediate the degradation of nitrated compounds by sulfides by at least two different pathways: reduction
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service