Skip to Content
Merck
All Photos(3)

Documents

765953

Sigma-Aldrich

1,11-Diazido-3,6,9-trioxaundecane

Synonym(s):

Azido-PEG3-azido, Bis-PEG3-Azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N6O3
CAS Number:
Molecular Weight:
244.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

refractive index

n20/D 1.470

bp

163-196 °C/760 mmHg

density

1.170 g/mL at 25 °C

storage temp.

−20°C

SMILES string

[N-]=[N+]=NCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C8H16N6O3/c9-13-11-1-3-15-5-7-17-8-6-16-4-2-12-14-10/h1-8H2

InChI key

SFMMXKLNFMIUCH-UHFFFAOYSA-N

Application

Homobifunctional PEG azide click chemistry linker. Azide functional groups will react via a copper catalyzed or strain promoted 1,3-dipolar cycloaddition click reaction with terminal alkynes and cyclooctyne derivatives to yield a stable triazole linkage.
1,11-Diazido-3,6,9-trioxaundecane may be used to synthesize:
  • 1,11-bis[4-(pyren-1-ylmethoxymethyl)-1H-1,2,3-triazole-1-yl]-3,6,9-trioxaundecane, a fluorogenic chemosensor that can selective detect Hg2+ and Ag+ ions in aqueous methanol solution
  • 1-amino-11-azido-3,6,9-trioxaundecane, an intermediate to synthesize tetraethylene glycol based bidentate ligand functionalized with dihydrolipoic acid and biotin (DHLA-TEG-biotin)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Selective Fluorescent Sensors for Hg2+ and Ag+ Based on Bis?triazole?Coupled Polyoxyethylenes in MeOH Solution.
Hung HC, et al.
European Journal of Organic Chemistry, 2009(36), 6360-6366 (2009)
Design of biotin-functionalized luminescent quantum dots.
Susumu K, et al.
BioMed Research International (2007)
Tilman Läppchen et al.
European journal of medicinal chemistry, 89, 279-295 (2014-12-03)
Calixarene 0118 is a potent anti-angiogenic agent that effectively inhibited tumor growth in preclinical studies, and is currently being evaluated in a phase I clinical trial. We have designed two close mimetics of calixarene 0118 containing a terminal alkynyl-functional group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service