735558
trans-1-Propenylboronic acid pinacol ester
97%
Synonym(s):
trans-1-Propeneboronic acid pinacol ester, trans-2-(1-Propen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.433
density
0.881 g/mL at 25 °C
SMILES string
C\C=C\B1OC(C)(C)C(C)(C)O1
InChI
1S/C9H17BO2/c1-6-7-10-11-8(2,3)9(4,5)12-10/h6-7H,1-5H3/b7-6+
InChI key
COPMASWDWLENMV-VOTSOKGWSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
127.9 °F
Flash Point(C)
53.3 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service