716839
Gold nanorods
10 nm diameter, λmax, 850 nm, dispersion in H2O
Synonym(s):
Au nanorods, Gold nanorod
About This Item
Recommended Products
form
colloidal suspension
dispersion in H2O
nanorod
Quality Level
contains
CTAB as stabilizer
concentration
>30 μg/mL in H2O
diam. × L
10 nm × 45 nm ± 10%
impurities
<0.1% CTAB
<0.1% ascorbic acid
diameter
10 nm
pH
3-5
density
1 g/mL at 25 °C
λmax
850 nm
Mw/Mn
(<10% CV, monodispersity)
>95 (rods)
storage temp.
2-8°C
SMILES string
[Au]
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Physical properties
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.
Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.
Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.
Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service