Skip to Content
Merck
All Photos(1)

Documents

713708

Sigma-Aldrich

(R)-(+)-2′-Amino-1,1′-binaphthalen-2-ol

97%

Synonym(s):

(R)-(+)-2-Amino-2′-hydroxy-1,1′-binaphthalene, (R)-NOBIN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO
CAS Number:
Molecular Weight:
285.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.5% (HPLC)
97%

form

crystals

optical purity

enantiomeric excess: ≥98.0%

InChI

1S/C20H15NO/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12,22H,21H2

InChI key

HIXQCPGXQVQHJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-2′-Amino-1,1′-binaphthalen-2-ol is a non-symmetrically substituted 1,1′-binaphthalene ligand.

Application

(R)-(+)-2′-Amino-1,1′-binaphthalen-2-ol may be used as a catalyst in the asymmetric PTC (phase-transfer catalysis) synthesis of α-amino acids. (R)-NOBIN reacts with trans-azobenzene-4,4′-dicarbonyl chloride to form poly(ester-amide)s, which shows backbone light-regulated chiroptical response.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Efficient Catalytic Synthesis of a-Amino Acids under Phase-Transfer Conditions with a Novel Catalyst/Substrate Pair.
Belokon YN, et al.
Angewandte Chemie (International Edition in English), 40(10), 1948-1951 (2001)
Stimuli-responsive polymers. 10. Photo-regulation of optical rotations in azobenzene modified poly (ester-amide) s containing highly structured, atropisomeric backbone geometries.
Lynch JG and Jaycox GD.
Polymer, 55(16), 3564-3572 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service