Skip to Content
Merck
All Photos(1)

Documents

665444

Sigma-Aldrich

6-Amino-1,3-dipropyluracil

97%

Synonym(s):

6-Amino-1,3-dipropyl-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N3O2
CAS Number:
Molecular Weight:
211.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

134-139 °C

SMILES string

CCCN1C(N)=CC(=O)N(CCC)C1=O

InChI

1S/C10H17N3O2/c1-3-5-12-8(11)7-9(14)13(6-4-2)10(12)15/h7H,3-6,11H2,1-2H3

InChI key

WWYIZMBRAYKRFU-UHFFFAOYSA-N

Application

6-Amino-1,3-dipropyluracil can be used in the synthesis of compounds of medicinal interest such as deazaflavins, pyrido[2,3-d]pyrimidines and tetrahydrobenzo[g]pyrimido[4,5-c]isoquinoline tetraones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferrocenyl, Alkyl, and Aryl?Pyrido [2, 3?d] Pyrimidines as Vasorelaxant of Smooth Muscle of Rat Aorta via cAMP Conservation Through Phosphodiesterase Inhibition.
Arellano I, et al.
Journal of Heterocyclic Chemistry, 53(4), 1147-1154 (2016)
Synthesis of Pyrimidine Fused Quinolines by Ligand-Free Copper-Catalyzed Domino Reactions.
Panday A K, et al.
The Journal of Organic Chemistry, 83(7), 3624-3632 (2018)
Cascade imination, Buchwald?Hartwig cross coupling and cycloaddition reaction: synthesis of pyrido [2, 3-d] pyrimidines.
Saikia P, et al.
Royal Society of Chemistry Advances, 5(30), 23210-23212 (2015)
2, 4-Dialkyl-8, 9, 10, 11-tetrahydrobenzo [g] pyrimido [4, 5-c] isoquinoline-1, 3, 7, 12 (2H, 4H)-tetraones as new leads against Mycobacterium tuberculosis.
Claes P, et al.
European Journal of Medicinal Chemistry, 77, 409-421 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service