652717
Poly(bis(4-carboxyphenoxy)phosphazene)
Synonym(s):
Poly(bis(carboxylatophenoxy)phosphazene)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
General description
Poly(bis(4-carboxyphenoxy)phosphazene) is a poly carboxylate that forms ionotropic hydrogel in the presence of metal cations.
Application
Poly(bis(4-carboxyphenoxy)phosphazene) may be used in preparing cell scaffolds, drug delivery, metal-sorbents and as substrates for microfluids.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shaped films of ionotropic hydrogels fabricated using templates of patterned paper.
Bracher P J, et al.
Advanced Materials, 21(4), 445-450 (2009)
Shaped films of ionotropic hydrogels fabricated using templates of patterned paper.
Bracher PJ, et al.
Advanced Mat., 21(4), 445-450 (2009)
Fabrication and manipulation of ionotropic hydrogels cross-linked by paramagnetic ions.
Winkleman A, et al.
Chemistry of Materials, 19(6), 1362-1368 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service