Skip to Content
Merck
All Photos(1)

Documents

62880

Sigma-Aldrich

L-Lysine ethyl ester dihydrochloride

≥99.0% (AT)

Synonym(s):

Lysine ethyl ester dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4CH(NH2)COOC2H5 · 2HCl
CAS Number:
Molecular Weight:
247.16
Beilstein:
3697099
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

solid

optical activity

[α]20/D +10±1°, c = 2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

Cl.Cl.CCOC(=O)[C@@H](N)CCCCN

InChI

1S/C8H18N2O2.2ClH/c1-2-12-8(11)7(10)5-3-4-6-9;;/h7H,2-6,9-10H2,1H3;2*1H/t7-;;/m0../s1

InChI key

DZIYAIZKJOHVQC-KLXURFKVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Vozzi et al.
Biomedical materials (Bristol, England), 13(5), 055006-055006 (2018-06-06)
Three-dimensional (3D) tissue models offer new tools in the study of diseases. In the case of the engineering of cardiac muscle, a realistic goal would be the design of a scaffold able to replicate the tissue-specific architecture, mechanical properties, and
Monica Boffito et al.
PloS one, 13(7), e0199896-e0199896 (2018-07-07)
Scaffolds populated with human cardiac progenitor cells (CPCs) represent a therapeutic opportunity for heart regeneration after myocardial infarction. In this work, square-grid scaffolds are prepared by melt-extrusion additive manufacturing from a polyurethane (PU), further subjected to plasma treatment for acrylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service