Skip to Content
Merck
All Photos(1)

Key Documents

603430

Sigma-Aldrich

Urea-13C

99 atom % 13C, UBT Grade

Synonym(s):

13C Labeled urea, Carbamide-13C, Carbonyldiamide-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N13CONH2
CAS Number:
Molecular Weight:
61.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

grade

cGMP

Quality Level

isotopic purity

99 atom % 13C

form

solid

mp

132-135 °C (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

N[13C](N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i1+1

InChI key

XSQUKJJJFZCRTK-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Analysis Note

Used as an Activie Pharmaceutical Ingredient (API) under the jurisdiction of the FDA and following cGMP and ICH guidelines for APIs
Used for Urea Breath Test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lotterer et al.
Zeitschrift fur Gastroenterologie, 29(11), 590-594 (1991-11-01)
A 13C-urea breath test for detection of Helicobacter pylori infection was validated in 306 patients. Breath samples (four, two or one) were taken at various time points within 30 minutes after a motility inhibiting liquid test meal with citric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service