Skip to Content
Merck
All Photos(1)

Documents

593052

Sigma-Aldrich

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

11 wt. %±5 wt. % in NMP/aromatic hydrocarbons (80%/20% solvent ratio)

Synonym(s):

Pyre-M.L.® RC-5097, Polyamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H12N2O .C10H2O6)x
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

concentration

11 wt. %±5 wt. % in NMP/aromatic hydrocarbons (80%/20% solvent ratio)

refractive index

n20/D 1.502

viscosity

105-135 poise

density

1.066 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Pyre-M.L. is a registered trademark of Industrial Summit Technologies Corp.

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingxian Yu et al.
Small (Weinheim an der Bergstrasse, Germany), 17(4), e2005925-e2005925 (2020-12-30)
Optical fibers made of polymeric materials possess high flexibility that can potentially integrate with flexible electronic devices to realize complex functions in biology and neurology. Here, a multichannel flexible device based on four individually addressable optical fibers transfer-printed with flexible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service