Skip to Content
Merck
All Photos(1)

Key Documents

576379

Sigma-Aldrich

Polyaniline (emeraldine base)

average Mw ~100,000

Synonym(s):

Emeraldine base polyaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mw ~100,000

Quality Level

conductivity

1 × 10-9 S/cm (pressed pellet, ASTM F8)

solubility

m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble

λmax

324 nm

SMILES string

Brc1n[nH]nc1C.Cl

InChI

1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H

InChI key

LYCWKZFKTMHXQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyaniline (PAni) emeraldine salt can be prepared by protonation of PAni emeraldine base (PAni-EB). PAni-EB forms at pH>7. Its mechanism against corrosive agents has been explained in a study.[1]
Polyaniline (emeraldine base) (PANI-EB) is an intrinsically conducting polymer (ICP) that can be formed by oxidative polymerization of aniline. Emeraldine base contains oxidized diimine and reduced diamine. PANI can be modified into the conjugating emeraldine base form by p-type doping with protonic acid.[2][3]

Application

PANI-EB can form a nanocomposite with multi-walled carbon nanotubes (MWCNTs) that can be used to functionalize glassy carbon electrodes for the electro-sensitive detection of cadmium (Cd(II)) and lead (Pb(II)).[3] Carbon nanofibers may be reinforced on PANI-EB to yield a specific capacitance of 95 F/g and a 92% retention over 3000 cycles as observed in supercapacitor applications.[4] It may be used with heptamethyleneimine as an inhibitor to form stable fuel cell membranes.[5]

Features and Benefits

Undoped form of conducting polymer (non-conductive).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of polyaniline membranes with-secondary amine additive containing N, N?-dimethyl propylene urea for fuel cell application
Kumar A, et al.
International Journal of Hydrogen Energy, 220, 267-275 (2018)
Engineering conformal nanoporous polyaniline via oxidative chemical vapor deposition and its potential application in supercapacitors
Li X, et al.
Chemical Engineering Science, 220, 267-275 (2018)
Polyanilines: a novel class of conducting polymers
MacDiarmid AG and Epstein AJ
Faraday Discussions of the Chemical Society, 88, 317-332 (1989)
Sensitive stripping voltammetric determination of Cd (II) and Pb (II) by a Bi/multi-walled carbon nanotube-emeraldine base polyaniline-Nafion composite modified glassy carbon electrode
Zhao G, et al.
Electrochimica Acta, 220, 267-275 (2016)
Study of epoxy and alkyd coatings modified with emeraldine base form of polyaniline.
Armelin E, et al.
Progress in Organic Coatings, 58, 316-322 (2007)

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service