Skip to Content
Merck
All Photos(1)

Key Documents

485969

Sigma-Aldrich

Boc-Phe-OH-1-13C

≥99 atom % 13C, ≥98% (CP)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-phenylalanine-1-13C, L-Phenylalanine-1-13C, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH-t-Boc)13CO2H
CAS Number:
Molecular Weight:
266.30
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥98% (CP)

form

solid

optical activity

[α]20/D +25°, c = 1 in ethanol

technique(s)

bio NMR: suitable

mp

85-87 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)[13C](O)=O

InChI

1S/C14H19NO4/c1-14(2,3)19-13(18)15-11(12(16)17)9-10-7-5-4-6-8-10/h4-8,11H,9H2,1-3H3,(H,15,18)(H,16,17)/t11-/m0/s1/i12+1

InChI key

ZYJPUMXJBDHSIF-GYZZUBJBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service