Skip to Content
Merck
All Photos(2)

Documents

483613

Sigma-Aldrich

Bis(trimethylstannyl)acetylene

Synonym(s):

1,2-Ethynediylbis[trimethylstannane], Ethynylenebis[trimethyl]tin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnC≡CSn(CH3)3
CAS Number:
Molecular Weight:
351.65
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

59-61 °C (lit.)

Quality Level

SMILES string

C[Sn](C)(C)C#C[Sn](C)(C)C

InChI

1S/C2.6CH3.2Sn/c1-2;;;;;;;;/h;6*1H3;;

InChI key

CDIFRACRLLNHOO-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service