Skip to Content
Merck
All Photos(1)

Key Documents

478962

Sigma-Aldrich

4-Bromobenzaldehyde diethyl acetal

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH(OC2H5)2
CAS Number:
Molecular Weight:
259.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.513 (lit.)

bp

75 °C/0.2 mmHg (lit.)

density

1.274 g/mL at 25 °C (lit.)

functional group

bromo
ether

SMILES string

CCOC(OCC)c1ccc(Br)cc1

InChI

1S/C11H15BrO2/c1-3-13-11(14-4-2)9-5-7-10(12)8-6-9/h5-8,11H,3-4H2,1-2H3

InChI key

BFSNEBVTOODGHZ-UHFFFAOYSA-N

General description

4-Bromobenzaldehyde diethyl acetal is also known as p-bromobenzaldehyde dimethyl acetal. It is employed during chemical decomposition of pladienolide D to benzylidene acetal.

Application

4-Bromobenzaldehyde diethyl acetal may be used in the preparation of 4-ethynylbenzaldehyde diethylacetal.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-ray crystal structures and conformational analysis of cyclic acetals derived from tartaric acid and rigid spacer units.
Ei?mann D, et al.
Structural Chemistry, 23(4), 1131-1142 (2012)
Total synthesis of the potent antitumor macrolides pladienolide B and D.
Kanada RM, et al.
Angewandte Chemie (International Edition in English), 46(23), 4350-4355 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service