Skip to Content
Merck
All Photos(4)

Documents

431559

Sigma-Aldrich

Lithium carbonate

greener alternative

99.99% trace metals basis

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

ACS reagent

Assay

99.99% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% insol. dil. HCl
≤0.2% S compounds
<100 ppm total metallic impurities

mp

618 °C (lit.)

density

2.11 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤5 ppm

cation traces

Ca: ≤0.01%
Fe: ≤0.002%
K: ≤0.01%
Na: ≤0.1%
heavy metals: ≤0.002%

greener alternative category

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Lithium carbonate - 99.99% trace metals basis, an inorganic compound with its chemical formula Li2CO3, is a white powder. It’s often used in ceramic and pharmaceutical, metallurgical industries etc. It is a key ingredient in alkaline storage battery, NMC111, NMC442, NMC532, NMC622 and LFP lithium-ion batteries.

Application

Lithium Carbonate is used in the field of high-energy lithium-ion batteries (automotive, energy storage) production, to produce materials such as LCO(Lithium Cobalt Oxide), LMO(Lithium ion Manganese Oxide), LTO(Lithium Titanate Oxide), LFP (Lithium iron phosphate), NMC111, NMC442, NMC532, NMC622 for Li-ion battery and those for other alkaline batteries. It is used to produce cathode materials such as lithium cobalt oxide (LiCoO₂) and lithium iron phosphate (LiFePO). Due to its crystalline stability Li2CO3 is widely used in industrial sectors, including pharmaceuticals, optical glass, ceramics, alloy, and lithium-ion batteries. Lithium carbonate can be used in research areas of bipolar disorder and other mental health conditions as a mood stabilizer. Lithium carbonate is used in the treatment of bipolar disorder and other mental health conditions as a mood stabilizer.

Features and Benefits

(1) Exceptional Purity: The 99.99% (<100 ppm) purity minimizes contamination from trace metals, ensuring suitability for applications sensitive to even minute impurities.

(2) Consistent Performance: Ultra-high purity guarantees consistent performance across various applications, reducing variability and enhancing reliability.

(3) High Purity Standard: Ideal as a standard or reagent for trace metal analysis and high-precision analytical techniques, ensuring accurate and reliable results.

(4) Lower conetent of anions such (Chloride and Nitrate)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siru Ren et al.
Journal of chromatography. A, 1609, 460519-460519 (2019-09-16)
In this work, tetraalkylammonium amino acid ionic liquids (TAA-AAILs) were first applied to non-aqueous capillary electrophoresis (NACE) to establish synergistic systems with a conventional chiral selector, native β-cyclodextrin (β-CD). Excellent enantioseparations of some dansyl-amino acid (Dns-AA) samples were achieved. A
Oxidative decomposition mechanisms of lithium carbonate on carbon substrates in lithium battery chemistries
Cao D, et al.
Nature Communications, 13, 4908-4908 (2022)
The Factors Influencing Lithium Carbonate Crystallization in Spent Lithium-Ion Battery Leachate
Gu K, et al.
Processes, 12(4), 753-753 (2024)
Conversion of CO2 to CO by electrolysis of molten lithium carbonate.
Kaplan V, et al.
Journal of the Electrochemical Society, 157(4), B552-B556 (2010)
Rodrigo Machado-Vieira et al.
Bipolar disorders, 11 Suppl 2(Suppl 2), 92-109 (2009-06-23)
Lithium has been and continues to be the mainstay of bipolar disorder (BD) pharmacotherapy for acute mood episodes, switch prevention, prophylactic treatment, and suicide prevention. Lithium is also the definitive proof-of-concept agent in BD, although it has recently been studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service