All Photos(1)
About This Item
Linear Formula:
BrC6H4CO2CH3
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
refractive index
n20/D 1.559 (lit.)
bp
252 °C (lit.)
density
1.532 g/mL at 25 °C (lit.)
functional group
bromo
ester
SMILES string
COC(=O)c1ccccc1Br
InChI
1S/C8H7BrO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,1H3
InChI key
SWGQITQOBPXVRC-UHFFFAOYSA-N
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service