Skip to Content
Merck
All Photos(1)

Key Documents

282316

Sigma-Aldrich

Perfluoro-1,3-dimethylcyclohexane

technical grade, 80%

Synonym(s):

Decafluoro-1,3-bis(trifluoromethyl)cyclohexane, Hexadecafluoro-1,3-dimethylcyclohexane, Hexadecafluoro-1,3-dimethylcyclohexane (cis+trans)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F10(CF3)2
CAS Number:
Molecular Weight:
400.06
Beilstein:
2064147
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

14 (vs air)

Assay

80%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

101-102 °C (lit.)

mp

−55 °C (lit.)

density

1.828 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(C(F)(F)F)C1(F)F

InChI

1S/C8F16/c9-1(7(19,20)21)3(11,12)2(10,8(22,23)24)5(15,16)6(17,18)4(1,13)14

InChI key

LOQGSOTUHASIHI-UHFFFAOYSA-N

General description

Quenching of the second excited singlet state of xanthione in inert, nonpolar perfluoro-1,3-dimethylcyclohexane has been studied using picosecond time-resolved fluorescence decay methods.

Application

Perfluoro-1,3-dimethylcyclohexane has been used in:
  • the preparation of amorphous fluorocarbon films by plasma polymerization
  • hydroformylation of linear terminal alkenes using rhodium based catalysts under fluorous biphasic conditions in the presence and absence of toluene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrical and optical properties of amorphous fluorocarbon films prepared by plasma polymerization of perfluoro-1, 3-dimethylcyclohexane.
Weber A, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 16(4), 2120-2124 (1998)
Meagan M Gadzuk-Shea et al.
The journal of physical chemistry. B, 122(43), 9947-9955 (2018-10-24)
Understanding the structures of proteins in the gas phase is essential for using gas-phase measurements to infer the properties of proteins in solution. Using serum albumin as a model, this study aims to expand our understanding of this relationship for
Hydroformylation in perfluorinated solvents; improved selectivity, catalyst retention and product separation.
<BIG>Foster DF, et al. </BIG>
Tetrahedron, 58(20), 3901-3910 (2002)
Transient effects in the fluorescence quenching of xanthione in perfluoro-1, 3-dimethylcyclohexane.
Maciejewski A, et al.
International Journal of Radiation Applications and Instrumentation, Part B: Nuclear Medicine and Biology, 39(1), 155-158 (1992)
Antonelle Pardo et al.
Journal of chromatography. A, 1364, 128-139 (2014-09-10)
Molecularly imprinted polymers (MIPs) based on quercetin and synthesized by either bulk, precipitation or suspension polymerization were characterized in terms of size and shape by scanning electron microscopy (SEM) and transmission electron microscopy (TEM). After a study of rebinding protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service