Skip to Content
Merck
All Photos(3)

Key Documents

269050

Sigma-Aldrich

5-Chloro-2-nitroaniline

97%

Synonym(s):

2-Amino-4-chloro-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NH2
CAS Number:
Molecular Weight:
172.57
Beilstein:
2210201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

chunks

mp

125-129 °C (dec.) (lit.)

functional group

chloro
nitro

SMILES string

Nc1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C6H5ClN2O2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H,8H2

InChI key

ZCWXYZBQDNFULS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The coupling of divinylbenzene cross-linked polystyrene with 5-chloro-2-nitroaniline followed by oxidative decyanation afforded benzophenone[1].

Application

5-Chloro-2-nitroaniline was used in the synthesis of 5-(4-substituted piperazin-1-yl)-2-nitroanilines and 5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates[2].

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Traceless solid-phase synthesis of 5-benzoylbenzimidazoles.
Canadian Journal of Chemistry, 79(11), 1556-1561 (2001)
R M Sánchez-Alonso et al.
Die Pharmazie, 44(9), 606-607 (1989-09-01)
A series of 5-(4-substituted piperazin-1-yl)-2-nitroanilines (4) and 5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates (6) has been synthesized starting from 5-chloro-2-nitroaniline (3) and N-monosubstituted piperazines. Catalytic reduction of 4 with Pd/C followed by treatment with 1,3-dicarbomethoxy-S-methylisothiourea yielded the corresponding methyl-5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates (6) which were for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service