Skip to Content
Merck
All Photos(1)

Key Documents

252670

Sigma-Aldrich

Chlorotriisopropoxytitanium(IV) solution

1.0 M in hexanes

Synonym(s):

Chlorotitanium(IV) triisopropoxide, Propan-2-olate titanium(4+) chloride, Titanium(IV) chloride triisopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]3TiCl
CAS Number:
Molecular Weight:
260.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solution

Quality Level

reaction suitability

core: titanium
reagent type: catalyst

concentration

1.0 M in hexanes

density

0.775 g/mL at 25 °C

SMILES string

CC(C)O[Ti](Cl)(OC(C)C)OC(C)C

InChI

1S/3C3H7O.ClH.Ti/c3*1-3(2)4;;/h3*3H,1-2H3;1H;/q3*-1;;+4/p-1

InChI key

IFMWVBVPVXRZHE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Chlorotriisopropoxytitanium(IV) solution may be used as a precursor for the synthesis of ultralow density ceramic materials. The product may be used to mediate a reductive amination reaction of 5α-cholestane-3,7-dione to yield aryl aminocholestanes.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation and properties of some σ-alkyl and σ-aryl derivatives of titanium.
Rausch MD and Gordon HB
Journal of Organometallic Chemistry, 74(1), 85-90 (1974)
Stereoselective synthesis of 3, 7-diarylaminocholestanes by titanium-mediated reductive amination.
Ahmad Md Wasi and Kim HS
Steroids, 88, 53-59 (2014)
Mesoporous Oxides and Their Applications to Hydrogen Storage
Hunt AJ, et al.
Material Matters, 4(2) null

Articles

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Sustainable energy sources with high production efficiency are crucial for meeting increasing energy demand.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service