243698
Rubidium hydroxide solution
50 wt. % in H2O, 99% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
RbOH
CAS Number:
Molecular Weight:
102.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
liquid
Recommended Products
grade
for analytical purposes
Assay
99% trace metals basis
form
liquid
concentration
50 wt. % in H2O
impurities
≤20000.0 ppm Trace Metal Analysis
density
1.74 g/mL at 25 °C
SMILES string
[OH-].[Rb+]
InChI
1S/H2O.Rb/h1H2;/q;+1/p-1
InChI key
CPRMKOQKXYSDML-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service